A propos de cet article
Autoclave-resistant blackout labels, designed for high-temperature as well as dry heat and steam sterilization applications.
- Temperature range: −60 to 150 °C (−76 to 302 °F)
- Unique opaque design allows labels to permanently obscure all pre-existing information
- Will withstand sterilization in steam autoclaves and dry heat ovens
- Compatible with all major thermal transfer barcode printers
- RR-class resin ribbon recommended
Labels have a strong permanent adhesive to ensure they won’t peel or wrinkle, even at elevated temperatures up to 150 °C (302 °F). These labels are compatible with a range of materials, including metal (stainless steel), glass (Pyrex), and plastic (Polypropylene), and can adhere strongly to both flat and curved surfaces.
The labels have a unique opaque design, allowing them to permanently block out previously printed information, including pre-existing text, serialized numbers, and barcodes. Thermal transfer printable, these blackout autoclave-resistant labels are ideal for bags and containers marked with sensitive or confidential information that must be autoclaved before disposal, such as biohazard and radioactive material.