PYREX® Distilling Flask, Vertical Type, Standard Taper 24/40 Joints, 3 Necks, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area. Three vertical neck flasks have two side necks with 24/40 outer standard taper joints and a center neck with a 24/40 standard taper joint. All the necks are at the same height.
Spécifications
- Catalog No:
- 89090-864
- 89090-696
- 89091-476
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey