VWR® Soft Nitrile Examination Gloves
Catalog # 89038-274
Supplier: VWR International
Specifications
- Glove Color:Blue
- Glove size:XL
- Ambidextrous/Hand-specific:Ambidextrous
- Disposable / reusable:Disposable
- Powdered/Powder-free:Powder-free
- Length:22.9 cm (9")
- Cuff thickness:2.3 mil
- Finger thickness:3.9 mil
- Palm thickness:2.7 mil
- Cat. no.:89038-274
Specifications
About this item
Ambidextrous gloves protect hands from a broad range of chemicals and biohazards.
- Ambidextrous
- Powder and latex free
- Textured
- Beaded cuffs
Ideal for all industrial and laboratory tasks.Made of premium compounded nitrile for tactile sensitivity and comfort.
Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins.
Customers Who Bought This Also Bought