ContactUsLinkComponent
Graduated Beakers with Handles, Stainless Steel, Polar Ware
Graduated Beakers with Handles, Stainless Steel, Polar Ware
# de catalogue 13983-067
Fournisseur:  Polar Ware
Fournisseur: T1063
Graduated Beakers with Handles, Stainless Steel, Polar Ware
Graduated Beakers with Handles, Stainless Steel, Polar Ware
# de catalogue 13983-067
Fournisseur:  Polar Ware
Fournisseur: T1063
Log in for availability
Select an Option
908 ml (30.7 oz.)
Select an Option
120 mm
Select an Option
133 mm
Select an Option
13983-067
Select an Option
T1063
A propos de cet article
Made of type 18-8 stainless steel and graduated in both ounces and mL
Tapered shape with spout and handle.
Customers Who Bought This Also Bought