GraLab® Darkroom Timer, Dimco-Gray
# de catalogue 62373-007
Fournisseur: Dimco-Gray
UL Listed
Spécifications
- Electrical:120 V, 60 Hz
- Supplier No.:7-300-160R
Spécifications
A propos de cet article
Ideal for photographic, graphic arts, and printing darkroom use.
- Provides 60 minutes of timing by minutes and seconds in one-second increments
- Buzzer alarm with variable volume control signals end of time period
Black face has a single circle of interval time numerals. Hands and numerals are phosphorescent. With two three-wire outlet receptacles. Chemical- and moisture-resistant case is made from a blend of hybrid plastics.
Certifications: UL listed.
Customers Who Bought This Also Bought