Esteem® Stretchy Nitrile Examination Gloves, Cardinal
Fournisseur: Cardinal Health
CE Compliant
Certificats
A propos de cet article
Powder-free nitrile gloves are ideal for applications requiring maximum tactility and sensitivity. Extra-stretchy nitrile formulation ensures a more comfortable fit than other nitrile gloves and conforms to hands quickly to facilitate a wide range of motion, especially around the thumb area. Gloves are puncture-resistant and have textured fingertips for a firm, no-slip grip. Beaded cuff resists tearing and minimizes cuff rolldown. Ambidextrous.
Textured gloves have a smooth surface and textured fingers.XP gloves have the same AQL as surgical gloves to ensure barrier protection.
Certifications: Gloves meet or exceed the bacteriophage penetration test as established by ASTM F1671.
Spécifications
- Catalog No:
- 76183-710
- 76183-734
- 76183-732
- 76183-708
- 76183-730
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free