Nalgene® Square Media Bottles with Septum Closure, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.
- Noncytotoxic
- Leakproof
- Sterile
- Offered in variety of sizes
The silicone/PTFE closure with septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity. Bottles and closures are sterile to 10 to 6 sterility assurance level.
Certifications: Biologically tested bottle and septum closure meet USP Class VI and European Pharmacopeia Abnormal Toxicity standards.
Ordering information: Bottles are packaged in shrink-wrapped trays in double poly-bagged cartons for cleanroom applications. Bottles are supplied two trays per case, except bottles 46600-586, which are supplied five trays per case, and bottles 46600-588, which are supplied four trays per case.
Frequently Bought Together