ContactUsLinkComponent
QuantiFluo™ α-Amylase Inhibitor Screening Kit
QuantiFluo™ α-Amylase Inhibitor Screening Kit
# de catalogue 77441-692
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
A-AMYLASE INHIBITOR SCREENING KIT
QuantiFluo™ α-Amylase Inhibitor Screening Kit
# de catalogue 77441-692
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiFluo™ α-Amylase inhibitor screening kit
  • Size:
    400 tests
  • Cat. no.:
    77441-692
Spécifications
A propos de cet article
For evaluation of drugs and screening potential inhibitors of α-amylase.
  • Safe
  • Non-radioactive assay
  • Fast and convenient
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be completed in under an hour at room temperature
  • Robust and High-throughput
  • A Z’-factor of >0.90 was observed in a 384-well format
  • Can be readily automated to assay thousands of samples per day
AMYLASE belongs to the family of glycoside hydrolase enzymes that break down starch into glucose molecules by acting on α-1,4-glycosidic bonds. The α-amylases (EC 3.2.1.1) cleave at random locations on the starch chain, ultimately yielding maltotriose and maltose, glucose, and 'limit dextrin' from amylose and amylopectin. In mammals, α-amylase is a major digestive enzyme. Increased enzyme levels in humans are associated with salivary trauma, mumps due to inflammation of the salivary glands, pancreatitis, and renal failure.
Simple, direct, and automation-ready procedures for measuring α-amylase inhibition are highly desirable in Research and Drug Discovery. BioAssay Systems’ QuantiFluo™ α-Amylase Inhibitor Screening Kit utilizes fluorescence polarization (FP) to screen for potential α-amylase inhibitors. In this assay, α-amylase cleaves a fluorescent amylose substrate. The decrease in FP is directly proportional to the α-amylase activity in the sample. Inhibition is therefore determined by the increase in FP (λex/em = 485/520 nm).
Customers Who Bought This Also Bought