ContactUsLinkComponent
QuantiFluo™ Urokinase Inhibitor Screening Assay Kit
QuantiFluo™ Urokinase Inhibitor Screening Assay Kit
# de catalogue 77441-672
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT UROKINASE INHIBITOR SCREENING
QuantiFluo™ Urokinase Inhibitor Screening Assay Kit
# de catalogue 77441-672
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiFluo™ Urokinase inhibitor screening assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-672
Spécifications
A propos de cet article
For evaluation of drugs and screening potential inhibitors of urokinase.
  • Safe
  • Non-radioactive assay
  • Fast
  • Assay is completed within a 30 minutes reaction time
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
UROKINASE PLASMINOGEN ACTIVATOR (urokinase, uPA) is a key serine protease involved in the degradation of the extracellular matrix that catalyzes the conversion of plasminogen to active plasmin. It acts as a thrombolytic agent to break up blood clots and when over-expressed, has been reported to influence the growth of certain malignant tumors (breast, prostate, etc.). BioAssay Systems’ DUKI-100 Kit provides a convenient fluorimetric means to screen for potential urokinase inhibitors. In this assay, the fluorimetric substrate reacts with urokinase and an inhibitor will decrease the fluorescence at λex/em = 380/450 nm.
Customers Who Bought This Also Bought