ContactUsLinkComponent
QuantiFluo™ Ammonia Assay Kit II
QuantiFluo™ Ammonia Assay Kit II
# de catalogue 77441-640
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT II AMMONIA
QuantiFluo™ Ammonia Assay Kit II
# de catalogue 77441-640
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiFluo™ Ammonia assay kit II
  • Size:
    100 tests
  • Cat. no.:
    77441-640
Spécifications
A propos de cet article
For quantitative determination of ammonia and ammonium in biological samples.
  • Fast and sensitive
  • Linear detection range of 0.005 to 2 mM ammonia
  • Convenient and high-throughput
  • A single component reagent
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
AMMONIA (NH₃) or its ion form ammonium (NH₄⁺) is an important source of nitrogen for living systems and is ubiquitously present in the nature. Simple, direct and automation-ready procedures for measuring NH₃ are very desirable. BioAssay Systems’ updated ammonia/ammonium assay is based on an improved o-phthalaldehyde method. This reagent reacts with ammonia/ammonium and forms a fluorescent product. The fluorescence intensity (λex/em = 415/475 nm) is proportional to the ammonia concentration in the sample.
Customers Who Bought This Also Bought