Spécifications
- Description:EnzyFluo™ farnesyltransferase inhibitor screening kit
- Size:400 tests
- Cat. no.:77441-686
Spécifications
A propos de cet article
For high-throughput screening of FTase inhibitors and evaluation of drug modulators.
- Safe and non-radioactive assay
- Homogeneous and convenient
- 'Mix-incubate-measure' type assay
- No wash and reagent transfer steps are involved
- High-throughput
- A Z - factor of 0.8 and higher is routinely observed in a 384-well format
- Can be readily automated to assay thousands of samples per day
Farnesyltransferase (FTase, EC 2.5.1.58) catalyzes the transfer of a farnesyl group from farnesyl pyrophosphate to the cysteine residue of the C-terminus of target proteins. When not properly regulated, farnesylated proteins, including the Ras superfamily of small GTPases, can lead to developmental disorders and cancer. Simple, direct and high-throughput inhibitor screening assays find wide applications for oncology research.
BioAssay Systems EIFT-400 assay kit provides a convenient fluorimetric method to screen for potential FTase inhibitors. In this assay, FTase reacts with farnesyl pyrophosphate and a dansyl-peptide substrate with measurable fluorescence at λem/ex = 340/550 nm. Inhibition is determined by the decrease in fluorescence.