ContactUsLinkComponent
EnzyChrom™ Formaldehyde Assay Kit
EnzyChrom™ Formaldehyde Assay Kit
# de catalogue 77441-682
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT FORMALDEHYDE
EnzyChrom™ Formaldehyde Assay Kit
# de catalogue 77441-682
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EnzyChrom™ formaldehyde assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-682
Spécifications
A propos de cet article
For formaldehyde determination in biological samples, food, beverages, and the environment.
  • Fast and sensitive
  • Use of 50 μl sample
  • Linear detection range 1.2 to 500 μM
  • Formaldehyde in 96-well plate assay
  • Convenient
  • The procedure involves adding a single working reagent and reading the absorbance after 30 minutes
  • Room temperature assay
  • High-throughput, 'Add-mix-read' type assay
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
Formaldehyde (methanal) is the simplest aldehyde. It is widely employed in industry for a wide range of applications. Formaldehyde is also used as a disinfectant and is a commonly utilized tissue fixative and embalming agent. Formaldehyde is naturally present in all tissues and body fluids. Recently, it has been shown that some cancer types exhibit elevated formaldehyde levels.
Increased formaldehyde concentration in urine has been associated with prostate and bladder cancer. Thus, measuring formaldehyde in urine can be a very useful tool when studying cancer. BioAssay systems formaldehyde assay kit is based on formaldehyde dehydrogenase catalyzed oxidation of formaldehyde, which generates formate and NADH that reduces a formazan (MTT) dye. The intensity of the reduced MTT, measured at 565 nm, is directly proportional to formaldehyde concentration in the sample.
Customers Who Bought This Also Bought