To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:EnzyChrom™ aldehyde dehydrogenase assay kit
- Size:100 tests
- Cat. no.:77441-678
Spécifications
A propos de cet article
For quantitative determination of aldehyde dehydrogenase activity in biological samples (e.g serum, cell lysate, etc).
- Sensitive and accurate
- Linear detection range 0.2 to 25 U/L ALDH in a 96-well plate assay
- Convenient
- The assay involves the addition of a single working reagent and can be completed in under an hour
- Robust and amenable to HTS
- Homogeneous 'mix-incubate-measure' type assay
- No wash and reagent transfer steps are involved
- Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
Aldehyde dehydrogenases (ALDHs) are a superfamily of oxidoreductases that catalyze the conversion of aldehydes to carboxylic acids. ALDH is crucial in the metabolism of alcohol as alcohol dehydrogenase breaks down ethanol to acetaldehyde.
Acetaldehyde, which is toxic to the body, is in turn broken down by ALDH to acetic acid. Imbalances of ALDHs have been linked to both alcoholism and alcohol sensitivity in people. BioAssay systems EADH-100 kit is based on the enzymatic conversion of acetaldehyde to acetic acid and NADH by ALDH.
The formed NADH is converted into a colored product, the absorbance of which, measured at 565 nm, is proportional to the enzyme activity in the reaction.