ContactUsLinkComponent
QuantiFluo™ Trypsin Inhibitor Assay Kit
QuantiFluo™ Trypsin Inhibitor Assay Kit
# de catalogue 77441-670
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT TRYPSIN INHIBITOR
QuantiFluo™ Trypsin Inhibitor Assay Kit
# de catalogue 77441-670
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiFluo™ trypsin inhibitor assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-670
Spécifications
A propos de cet article
For evaluation of drugs and screening of potential inhibitors to trypsin proteases.
  • Safe and non-radioactive assay
  • Homogeneous and convenient
  • Homogenous 'mix-incubate-measure' type assay
  • No wash and reagent transfer steps are involved
  • Robust and high-throughput
  • Can be readily automated to assay thousands of samples per day
  • Robust assay with a Z factor of >0.7
Trypsin (EC 3.4.21.4) is a digestive, serine protease that hydrolyzes dietary proteins in many eukaryotic and prokaryotic organisms. Trypsin predominantly cleaves peptide chains at the carboxyl side of lysine and arginine amino acids, but not before proline. BioAssay systems QuantiFluo™ trypsin inhibitor assay kit uses a fluorescein isothiocyanate (FITC)-labeled synthetic substrate.
The fluorescein label is highly quenched. Upon digestion by trypsin present in the sample, the substrate is cleaved into smaller peptides, which abolishes the quenching of the fluorescence label. The fluorescence or fluorescence polarization (FP) of the FITC-labeled fragments is measured at λex/em = 485/530 nm. Inhibition is determined by the decrease in fluorescence.
Customers Who Bought This Also Bought