ContactUsLinkComponent
QuantiChrom™ Pectinase Assay Kit
QuantiChrom™ Pectinase Assay Kit
# de catalogue 77441-662
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT PECTINASE
QuantiChrom™ Pectinase Assay Kit
# de catalogue 77441-662
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiChrom™ pectinase assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-662
Spécifications
A propos de cet article
For quantitative determination of pectinase activity determination in biological samples.
  • Linear detection range from 9.2 to 100 U/L pectinase activity in a 96-well plate assay
  • Simple and convenient 40 - minute 'add-mix-measure' type assay
  • High-throughput format and compatible with laboratory liquid handling systems
  • No heating is required
  • The kit does not use toxic materials
  • Refer to the previously established DNS method
Pectinase (EC 3.2.1.15) is an enzyme that catalyzes the hydrolysis of pectin, a heteropolysaccharide found primarily in the middle lamella and cell wall of terrestrial plants. During fruit ripening, enzymes, including pectinase break down the middle lamella, separating cells and softening the fruit. Pectinase enzymes are used in the food industry to liquefy fruit pulp and clarify fruit juice and wine.
BioAssay systems DPEC-100 kit provides a rapid and convenient assay for pectinase enzyme activity, in which pectin the substrate forms a turbid complex with the detection reagent. The turbidity, measured at OD600 nm, is proportional to the amount of unhydrolyzed pectin, thus inversely proportional to the pectinase activity.
Customers Who Bought This Also Bought