ContactUsLinkComponent
QuantiChrom™ Cobalt Assay Kit
QuantiChrom™ Cobalt Assay Kit
# de catalogue 77441-654
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT COBALT
QuantiChrom™ Cobalt Assay Kit
# de catalogue 77441-654
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiChrom™ cobalt assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-654
Spécifications
A propos de cet article
Direct assays: Cobalt in aqueous samples.
  • Sensitive and accurate
  • Linear detection range 1.4 μM to 200 μM cobalt in 96-well plate assay
  • Simple and high-throughput
  • The procedure involves addition of a single working reagent with immediate color development
  • Can be readily automated as a high-throughput assay for thousands of samples per day
  • Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent stability
  • Cuvette or 96-well plate assay
Cobalt is an essential micronutrient for all multicellular organisms as the active center of cobalamins, such vitamin B-12, which is essential for plants and animals. Cobalt is also a micronutrient for bacteria, algae, and fungi. Despite being an important micronutrient, excess cobalt in the environment can be deleterious to life leading to cardiomyopathy in humans and necrosis in plants.
Simple, direct and automation-ready procedures for measuring cobalt concentrations find wide applications in research and environmental monitoring. BioAssay systems cobalt assay kit is designed to measure total cobalt directly in aqueous samples without any pretreatment. The intensity of the color, measured at 485 nm, is directly proportional to the cobalt concentration in the sample.
Customers Who Bought This Also Bought