ContactUsLinkComponent
QuantiChrom™ Acetoacetate Assay Kit
QuantiChrom™ Acetoacetate Assay Kit
# de catalogue 77441-642
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
ASSAY KIT ACETOACETATE
QuantiChrom™ Acetoacetate Assay Kit
# de catalogue 77441-642
Fournisseur:  BIOASSAY SYSTEMS
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiChrom™ acetoacetate assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-642
Spécifications
A propos de cet article
Direct assay of acetoacetate in serum, plasma, urine and other biological samples.
  • Sensitive and accurate
  • Linear detection range of 0.012 to 1 mM (1.2 to 100 nmole/well) for acetoacetate in 96-well plate assay
  • Convenient
  • No sample pretreatment needed
  • The procedure involves adding a single working reagent and reading the optical density at room temperature in kinetic mode
  • High-throughput
  • Can be automated as a high-throughput 96-well plate assay for thousands of samples per day
Acetoacetate is one of three ketone bodies produced in the liver mainly from oxidation of fatty acids and are normally present at low concentrations in urine and blood. Increased ketone concentrations in the blood may lead to metabolic acidosis, which has been associated with diabetes, childhood hypoglycemia, growth hormone deficiency, alcohol or salicylate intoxication and inborn errors of metabolism.
Acetoacetate is an unstable ketone body that spontaneously decarboxylates to release carbon dioxide and acetone. Simple, direct and automation-ready procedures for measuring acetoacetate (AAT) are very desirable. BioAssay Systems QuantiChrom™ acetoacetate assay is based on a reaction with sodium nitroprusside forming a product with absorbance at 550 nm.
Customers Who Bought This Also Bought