BioSoft™ Disposable Bio-Based Foam Ear Plugs with Pinchfit™ Technology
Fournisseur: PIP
New Product
Certificats
A propos de cet article
PIP® understands the importance of reducing waste to lower costs and minimize environmental impact. The new, sustainable ECOSeries™ BioSoft™ ear plugs are the first to be made from bio-based technologies.
- Soft, smooth, low pressure for excellent comfort
- Bio-based materials made in North America
- Independently tested NRR 32/SNR 38
- When stored in original packaging, BioSoft™ maintains the same shelf life as standard PU foam ear plugs
The BioSoft™ ear plug foam material provides the same fit and performance as conventional polyurethane or PVC materials, but with a lower carbon footprint. This innovative technology not only reduces emissions during manufacturing, but it contains bio-based materials making BioSoft™ more environmentally friendly at end of use. They are also recyclable through PIP's Terracycle program.
Customers Who Bought This Also Bought