Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Plain Bottom, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Graduated flasks are made of light, crystal-clear PETG plastic with easy-to-open polyethylene screw closures.
- Disposing of flasks reduces chance of cross-contamination
- Non-pyrogenic, non-cytotoxic
- Sterilized to SAL 10-6
The leak-proof high-density polyethylene screw closures open to vent with 1/4 turn.
Ideal for shaker and suspension cell culture, media preparation, or storage
Packaging: Individually packaged for easy storage.
Customers Who Bought This Also Bought