BioX Multi-Well Cell Culture Plates, TC Treated
Fournisseur: TWD Scientific
New Product
Certificats
A propos de cet article
Ideal for the cultivation and observation of sensitive cells.
- TC treated
- Highly transparent polystyrene
- Stackable design
- DNase/RNase free, non-pyrogenic
- Flat bottom
Treated polystyrene surface is great for both adhesion and extension of cells.
Customers Who Bought This Also Bought