KIMAX® Engler Distilling Flasks, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Sidearm tube is sealed at an angle of 75° from the neck and is 137 ± 3 mm from the bottom of the flask.
26016-125 version has 3 reference lines and is for use with Haage automatic distillation apparatus or others that call for three thermometer depth insertions.
Intended for ASTM D86, D233, D801, and D802.
Customers Who Bought This Also Bought