SP Wilmad-LabGlass Church Distilling Flasks, SP Industries
# de catalogue 14202-192
Fournisseur: SP Industries
Spécifications
- Capacity:300 ml
- Height:130 mm
- Cat. no.:14202-192
- Rubber Stopper Size:4
- Supplier No.:LG-7228-100
Spécifications
A propos de cet article
Specified for the distillation of bituminous samples, these borosilicate glass Church Distilling Flasks have a 10 mm I.D
sidearm and a neck made to fit a rubber stopper.
Certifications: Reference: ASTM D 20 Test Method for Distillation of Road Tars. ASTM D 246 Method for Distillation of Creosote and Creosote Coal Tar Solutions. ASTM D 402 Test Method for Distillation of Cut-Back Asphaltic (Bituminous) Products. ASTM D 2569 Test Method for Distillation of Pitch.
Customers Who Bought This Also Bought