KIMBLE® Barrett Distilling Flasks, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These flasks feature smooth, consistent, heavy walls for uniform heating and minimal breakage.
- Designed to ASTM E133 specifications
- Manufactured from 33 expansion, low extractable borosilicate glass
610910-0125 conforms to Flask B in ASTM E133.
610900-0200 conforms to Flask C in ASTM E133 and also is compatible for use with ASTM D1078.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Flask is supplied with one cork for the sidearm.
Customers Who Bought This Also Bought