Powder-Free Sterile Nitrile Gloves, CT International
Fournisseur: CT International
Certificats
A propos de cet article
NSGF series are powder free class 100 nitrile gloves.
- Like all our gloves, they are manufactured for outstanding strength, stretch and fit
They offer a textured surface and are double chlorinated reducing contact sensitizers and protein content.
Like all our gloves, they are manufactured for outstanding strength, stretch and fit.
Certifications: NSGF series class 100 nitrile gloves ISO 5 clean room compatible or higher for controlled environments.
Spécifications
- Catalog No:
- 56667-072
- 56667-074
- 56667-076
- 56667-078
- 56667-080
- 56667-082
- 56667-084
- 56667-070
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- FDA regulated:
- +
- +
- +
- +
- +
- +
- +
- ASTM No.:
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05