TouchNTuff® 69-318 Natural Rubber Latex Disposable Gloves, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
SType 69-318 gloves are 100% natural rubber latex.
- Powder-free
- Ambidextrous
- Textured grip
Gloves are suitable for laboratory analysis/technical work, food processing and handling, quality control, electronics assembly, as well as light-duty maintenance and cleanup.
Certifications: Gloves qualify as Class I Medical Devices. Component materials comply with FDA regulations for food contact.
Caution: This product contains natural rubber latex which may cause allergic reactions in some individuals.
Spécifications
- Catalog No:
- 36888-216
- 36888-210
- 36888-214
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powdered
- Powder-free