VWR® Traceable® Micro-Anemometer with Thermometer
# de catalogue 89094-736
Fournisseur: VWR International
Spécifications
- Description:Anemometer with Thermometer
- Speed resolution:0.1 m/sec., 0.1 ft./min., 0.1 mph, 0.1 km/hour, 0.1 knots
- Temperature accuracy:±1°C (±1.8°F)
- Temperature resolution:0.1°
- Temperature range:–20 to 80°C (–4 to 176°F)
- Dimensions:4.5 x 10.2 x 1.6 cm (13/4 x 4 x 5/8")
- Speed accuracy:±5% full scale
- Speed range:0 to 30 m/sec., 0 to 98 ft./sec., 0 to 67 mph, 0 to 108 km/hour, 0 to 58 knots
- Weight:28.4 g (1 oz.)
- Cat. no.:89094-736
Spécifications
A propos de cet article
Compact anemometer displays air velocity in meters per second, feet per second, miles per hour, kilometers per hour, and knots.
- Traceable®
- Displays air velocity, max. air velocity measured, temperature, wind chill temperature
- Tough ABS plastic housing
- Temperature in °C and °F
Unit also displays maximum air velocity measured, temperature, and wind chill temperature. Temperature is displayed in °C and °F. Measurements are updated twice per second. Tough ABS plastic housing and water-resistant design keep out moisture, dust, and dirt, assuring long life in severe lab or harsh plant environments. Features include intuitive one-button operation and electroluminescent backlight. High-contrast LCD digits are 1 cm (3/8") high.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Customers Who Bought This Also Bought