KIMAX® Babcock Bottle, Ice Cream and Half and Half Tests, 20%, Kimble Chase
# de catalogue 48720-009
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Height:165 mm (61/2")
- Tolerance:0.002%
- Sample Size:9 g (0.32 oz.)
- Supplier No.:516-20
Spécifications
A propos de cet article
KIMAX® bottle is filled through the graduated neck.
Scale is permanent brown stain. Designed for use in APHA test procedures.
Customers Who Bought This Also Bought