Dispensing Cannulas for SOCOREX® DOSYS™ Syringes, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Blunt end dispensing cannulas with Luer Lock. Ideal for use with 24 and 96 well microplates.
- Stainless steel
- Autoclavable
This unique dispensing system enables you to perform sample inoculations and cell feedings faster and more efficiently under sterile conditions.
Customers Who Bought This Also Bought