KIMAX® Babcock Bottle, Cream and Cheese Test, 50%, Sealed, Kimble Chase
Catalogue # 48712-007
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Graduation Interval:0.5% (0.1 ml)
- Height:165 mm (6.5")
- Size:50% (10 ml)
- Cat. no.:48712-007
- Tolerance:±0.5%
- Sample Size:18 g (0.64 oz.)
- Supplier No.:2015S-50
Spécifications
À propos de cet article
Designed for testing butter fat content.
- Scale is permanent brown stain
Neck is graduated to 50% in 0.5% subdivisions. Marked 'Sealed 3' for use in states requiring this special marking.
Certifications: Made in accordance with the specifications of the American Dairy Science Association, and the AOAC®/APHA.