ContactUsLinkComponent
KIMAX® Babcock Bottle, Milk Test, 8%, Sealed, Kimble Chase
Catalogue # 48711-056
undefined
KIMAX® Babcock Bottle, Milk Test, 8%, Sealed, Kimble Chase
Catalogue # 48711-056
Supplier Number:  1003S-8
Spécifications
  • Height:
    165 mm (6½")
  • Cat. no.:
    48711-056
  • Tolerance:
    ±0.05%
  • Sample Size:
    18 g (0.63 oz.)
  • Supplier No.:
    1003S-8
Spécifications
À propos de cet article
Designed for use in AOAC®/APHA test procedures.
Neck is graduated to 8% in 0.1% subdivisions. Etched vertical line, used to anchor calipers is centered in a contrasting darkened band which makes visualization easier. Marked with 'Sealed 3' for use in states requiring this marking.