Safegrip® Powder-Free Latex Gloves, Microflex®, Ansell
Fournisseur: Microflex
CE Compliant
Certificats
A propos de cet article
These gloves are up to four times thicker than standard examination gloves.
- Offering added durability in the palm and fingers
- Thickness: 16.5 mil (finger)
Ambidextrous gloves are fully textured to provide a superior grip in wet and dry conditions.
Packaging: Packaged 50 gloves per dispenser box.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex sensitized individuals has not been established.
Spécifications
- Catalog No:
- CA32916-600
- CA32916-604
- CA32916-602
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- Finger thickness:
- 16.5 mil
- 16.5 mil
- 16.5 mil
- Palm thickness:
- 11.4 mil
- 11.4 mil
- 11.4 mil