FLEXAPRENE* GREEN Powder-Free Examination Gloves
Fournisseur: O&M HALYARD, INC
New Product
Certificats
A propos de cet article
The FLEXAPRENE* GREEN exam glove is an advanced chloroprene technology that provides the feel and tactile sensitivity of latex without the risk of type 1 allergic reactions to natural rubber latex proteins. Additionally, these gloves offer the protection expected from a synthetic.
- 9.5" length
- Ambidextrous
- Tactile sensitivty comparable to latex
- Soft to the touch, yet powerful in grip
- Polymer coating for improved donning
- Not made with natural rubber latex
- Powder-free chloroprene technology
Can an exam glove really be that comfortable? With the FLEXAPRENE* GREEN exam glove, the answer is yes. Advanced chloroprene technology gives them the feel and tactile sensitivity of latex without the risk of Type I allergic reactions to natural rubber latex proteins. They are soft, easy to don, and fit so comfortably that wearers may forget to take them off.
Spécifications
- Catalog No:
- 77441-630
- 77441-638
- 77441-636
- 77441-634
- 77441-632
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Chloroprene
- Chloroprene
- Chloroprene
- Chloroprene
- Chloroprene
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile