To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ZNF423
- Clonality:Polyclonal
- Gene ID:23090
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:145
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Spécifications
A propos de cet article
Transcription factor that can both act as an activator or a repressor depending on the context. Plays a central role in BMP signaling and olfactory neurogenesis. Associates with SMADs in response to BMP2 leading to activate transcription of BMP target genes. Acts as a transcriptional repressor via its interaction with EBF1, a transcription factor involved in terminal olfactory receptor neurons differentiation; this interaction preventing EBF1 to bind DNA and activate olfactory-specific genes. Involved in olfactory neurogenesis by participating in a developmental switch that regulates the transition from differentiation to maturation in olfactory receptor neurons. Controls proliferation and differentiation of neural precursors in cerebellar vermis formation.
Type: Primary
Antigen: ZNF423
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat