Order Entry
ContactUsLinkComponent
Anti-Acetyl Coenzyme A Carboxylase beta Ser220 Rabbit Polyclonal Antibody
Anti-Acetyl Coenzyme A Carboxylase beta Ser220 Rabbit Polyclonal Antibody
# de catalogue 77440-402
Fournisseur:  Bioss
New Product
Anti-Acetyl Coenzyme A Carboxylase beta Ser220 Rabbit Polyclonal Antibody
# de catalogue 77440-402
Fournisseur:  Bioss
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Acetyl Coenzyme A Carboxylase beta Ser220
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µl
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    265
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Purified by protein A
  • Packaging:
    Vial

Spécifications

A propos de cet article

Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene. [provided by RefSeq, Jul 2008].

Type: Primary
Antigen: Acetyl Coenzyme A Carboxylase beta Ser220
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity:

Customers Who Bought This Also Bought