To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CSFV E2 Protein
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Virus
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:42
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Packaging:Vial
Spécifications
A propos de cet article
E(rns), E1 and E2 are responsible of cell attachment and subsequent fusion of viral and cellular membrane. P7 forms a leader sequence to properly orient NS2 in the membrane.
Uncleaved NS2-3 is required for production of infectious virus. NS2 protease seems to play a vital role in viral RNA replication control and in the pathogenicity of the virus. NS3 displays three enzymatic activities: serine protease, NTPase and RNA helicase. NS4A is a cofactor for the NS3 protease activity. RNA-directed RNA polymerase NS5 replicates the viral (+) and (-) genome.
Type: Primary
Antigen: CSFV E2 Protein
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Virus