To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:LXR alpha + LXR beta
- Clonality:Polyclonal
- Gene ID:10062
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Xenopus laevis,Mouse
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:51
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Packaging:Vial
Spécifications
A propos de cet article
LXR alpha and LXR beta are orphan receptors. Interaction with RXR shifts RXR from its role as a silent DNA binding partner to an active ligand binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4 type response elements characterized by direct repeats of two similar hexanuclotide half sites spaced by four nucleotides. Plays an important role in the regulation of cholesterol homeostasis. LXR alpha: Visceral organs specific expression. Strong expression was found in liver, kidney and intestine followed by spleen and to a lesser extent the adrenals. LXR beta: Ubiquitously expressed.
LXR alpha: Visceral organs specific expression. Strong expression was found in liver, kidney and intestine followed by spleen and to a lesser extent the adrenals. LXR beta: Ubiquitously expressed.
Type: Primary
Antigen: LXR alpha + LXR beta
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Xenopus laevis