KIMBLE® Gas Washing Bottles, with Fritted Cylinder, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Tall form bottles feature a coarse or extra coarse porosity fritted cylinder to maximize dispersion, 29/42 standard taper joints.
- 2 hose connectors
- Manufactured from 33 expansion, low extractable borosilicate glass
Supplied complete with hooks and two 5.1 cm (2") springs.
Unit contains a fritted disc for more uniform gas distribution throughout the absorbing material.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought