Ammonia Standard, 1 ml = 1 mg NH₃, 1000 ppm NH₃ (822 ppm N), Ricca Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
1000ppm as NH3, 822ppm nitrogen.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.