Active PoE Splitter, Direct Power and Data Splitting for POE-Enabled SenseAnywhere AccessPoints
# de catalogue 77441-696
Fournisseur: SENSEANYWHERE BV
New Product
Spécifications
- Description:Active PoE splitter: Direct power and data splitting for POE-enabled SenseAnywhere AccessPoints
Spécifications
A propos de cet article
Streamline the SenseAnywhere AccessPoint setup with our active POE splitter. Ideal for POE-enabled switch environments, it simplifies connection by splitting both power and data directly, eliminating the need for separate power supplies. A compact, efficient solution for seamless integration.
- Active Power Over Ethernet (POE) splitter: Simplifies the connection process by splitting power and data from a single cable into separate inputs for the AccessPoint
- Designed for POE-Enabled switches: Ideal for environments where the switch already provides both power and data, requiring only splitting at the device end
- Streamlines installation: Eliminates the need for separate power supplies, making installation cleaner and more efficient
- Universal compatibility: Works seamlessly with SenseAnywhere AccessPoints, ensuring reliable and continuous power and data connectivity
When the customer has a POE enabled switch the switch does provide both power and data and users only need to split them using the black active POE splitter before entering the AccessPoint.
Customers Who Bought This Also Bought