Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
# de catalogue 16354-320
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:30 ml (1 oz.)
- Cat. no.:16354-320
- Supplier No.:2414-0030
Spécifications
A propos de cet article
Made of Teflon® FEP and ETFE dropping closure and cap. Transparent bottle can be used with virtually any chemical. Contamination-free dispensing. Repeatedly autoclavable with tip uncapped.
Convenient one-hand operationjust squeeze bottle to dispense. Dispenses one drop at a time.
Customers Who Bought This Also Bought