Nalgene® Dispensing Bottles, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Produce a fine steady stream but only when squeezed. Linerless, noncontaminating 24-415 closure with captive spout cap. Leakproof. Bottles may be autoclaved full, with tips uncapped.
For dispensing sterile solutions.
Customers Who Bought This Also Bought