Nalgene® Drop-Dispenser Bottles, Low-Density Polyethylene, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Bottles dispense one drop at a time.
Captive spout cap won't release or leak when bottle is squeezed.
Delivery: Supplied with polypropylene dropping closure and spout plug.
Customers Who Bought This Also Bought