Nunc™ Cryobank™ Dense Storage Tubes and Cryoboxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Thermo Scientific Dense Low Temperature Storage format uses standard CryoBox sizes that work with the freezer racks you currently own
These dense storage tubes and cryoboxes are made of polycarbonate.
By using standard boxes with 13 x 13 tube separators, 169 1mL tubes fit in a standard size CryoBox – effectively doubling your capacity, and cutting storage costs by 50%.
Specifications
- Catalog No:
- 55710-180
- CA30618-016
- CA89238-098
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)