ContactUsLinkComponent
Chemware® Beakers, Teflon® PFA, Saint-Gobain Performance Plastics
Catalogue # 13917-584
Fournisseur #: D1069538
undefined
Chemware® Beakers, Teflon® PFA, Saint-Gobain Performance Plastics
Catalogue # 13917-584
Supplier Number:  D1069536
Fournisseur #: D1069538
S'identifier pour la disponibilité
Select an Option
50 - 250 ml
Select an Option
250 ml (8.5 oz.)
Select an Option
13917-584
Select an Option
D1069538
À propos de cet article
These low metal ion content PFA beakers are ideal for trace metal analysis and ultra-pure applications.
  • Special machined bottom design for rapid heat transfer
  • Transparent for easy viewing
  • Graduated for easy measuring
  • Non-stick, easy to clean surface
Microwave heatable, or heat directly on hot plate up to 500°F (260°C) when proper care is exercised.
Frequently Bought Together