ContactUsLinkComponent
Microtube Rack, 80-Well
Microtube Rack, 80-Well
# de catalogue 77395-444
Fournisseur:  Heathrow Scientific
New Product
RACK MICROTUBE 80-WELL RED PK5
Microtube Rack, 80-Well
# de catalogue 77395-444
Fournisseur:  Heathrow Scientific
New Product
Spécifications
  • Color:
    Red
  • Material:
    PP
  • For Tubes:
    1.5/2.0 ml tubes, 80 wells (11 mm well Ø)
Spécifications
A propos de cet article
Popular fraction-collector-plate style rack.
  • Compact 5×16 array makes filling and transfer easy
  • Frosted labeling area on bottom of rack
  • Embossed index numbers for quick sample ID and retrieval
  • Made of polypropylene
Easily fill and transfer 80 tubes in a compact arrangement with the 80-well microtube rack.
Customers Who Bought This Also Bought