Microtube Rack, 80-Well
# de catalogue 77395-444
Fournisseur: Heathrow Scientific
New Product
Spécifications
- Color:Red
- Material:PP
- For Tubes:1.5/2.0 ml tubes, 80 wells (11 mm well Ø)
Spécifications
A propos de cet article
Popular fraction-collector-plate style rack.
- Compact 5×16 array makes filling and transfer easy
- Frosted labeling area on bottom of rack
- Embossed index numbers for quick sample ID and retrieval
- Made of polypropylene
Easily fill and transfer 80 tubes in a compact arrangement with the 80-well microtube rack.
Customers Who Bought This Also Bought