Chemware® Griffin Beakers, PTFE, Saint-Gobain Performance Plastics
# de catalogue 13914-290
Fournisseur: Saint Gobain Performance Plastics
Fournisseur: D1069041
Log in for availability
A propos de cet article
These fluoropolymer beakers are non-contaminating and completely inert to chemical attack.
- Unbreakable
- Easy to clean
- Lightweight
- Microwavable
Beakers can be heated on a hot plate up to 288°C (550°F) when the temperature is monitored. Specially molded bottom allows rapid heat transfer. Broad spout for easy pouring.
Customers Who Bought This Also Bought