Nalgene® Fluorinated High-Density Polyethylene Jerricans, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Fluorocarbon surfaces inside and out reduce solvent absorption and penetration.
- Leakproof
Excellent for use with most acids, alkalies, and aggressive organic solvents. Long, integral spout allows for easy pouring. Molded-in graduations in liters and gallons.
Ordering information: Includes 53B fluorinated polypropylene closures.
Customers Who Bought This Also Bought