ContactUsLinkComponent
Phlebotomy Tray Lid, White
Phlebotomy Tray Lid, White
# de catalogue 77395-366
Fournisseur:  VWR International
New Product
COVER DROPLET PHLEBOTOMY TRAY WHT
Phlebotomy Tray Lid, White
# de catalogue 77395-366
Fournisseur:  VWR International
New Product
Spécifications
  • Description:
    Phlebotomy tray cover/lid, PS, white, for Phlebotomy/sample collection tray 77395-358 or 77395-360
  • Cat. no.:
    77395-366
  • L×W×H:
    420×240×205 mm (16.5×9.4×8")
Spécifications
A propos de cet article
Protect the contents of the Phlebotomy tray with this Phlebotomy tray lid. The custom designed sturdy lid is a snug fit, protects contents and provides access to the tray handle in its vertical position.
  • Made of polystyrene
  • Can be secured with cable ties for extra security
  • Stackable
Customers Who Bought This Also Bought