Dairy Jugs, High-Density Polyethylene, Qorpak®
Fournisseur: Avantor
Certificats
A propos de cet article
Natural HDPE Dairy Jugs are lightweight and are ideal for sampling and storeroom use.
- Less expensive
These general purpose containers provide an excellent moisture barrier and are impact resistant.
Customers Who Bought This Also Bought