Double-Ended Applicator Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Two-sided applicator brushes. On one side, the brushes have a short flat end, and on the other side they have an extra stiff chiseled end.
- Anti-static
- Fibers are mechanically clamped into the handle, eliminating adhesives and ensuring maximum fiber retention
- Great alternative to hand-trimming acid brushes
Chiseled end reaches and scrubs tight corners. Short, flat end is ideal for heavy-duty scrubbing.
Customers Who Bought This Also Bought